missing translation for 'onlineSavingsMsg'
Learn More

n-Pentane For gas chromatography ECD and FID SupraSolv™, MilliporeSigma™

Catalog No. M1008822500
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1008822500 2.5 L Glass Bottle
M1008821000 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1008822500 Supplier Technidata Supplier No. 1.00882.2500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For gas chromatography ECD and FID SupraSolv™

Chemical Identifiers

CAS 109-66-0
Molecular Formula C5H12
Molecular Weight (g/mol) 72.15
MDL Number MFCD00009498
InChI Key OFBQJSOFQDEBGM-UHFFFAOYSA-N
Synonym n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish
PubChem CID 8003
ChEBI CHEBI:37830
IUPAC Name pentane
SMILES CCCCC

Specifications

Melting Point -130°C
Color Colorless
Density 0.63g/cm3 at 20°C
Quantity 2.5 L
Flash Point 48°C (54°F)
Ignition Point 285°C (545°F)
Odor Benzine-like
Packaging Glass Bottle
Physical Form Liquid
Vapor Pressure 570 hPa (20°C)
UN Number 1265
Solubility Information Soluble in water: 0.4g/L at 16°C
Boiling Point 36°C
Formula Weight 72.15
Chemical Name or Material n-Pentane
Show More Show Less
Hazard Category H225: Highly flammable liquid and vapour.
Hazard Statement H304: May be fatal if swallowed and enters airways.
Precautionary Statement H336 May cause drowsiness and dizziness.
Recommended Storage 5°C to 30°C (41°F to 86°F); Keep container tightly closed in a dry and well-ventilated place. Keep away from heat and sources of ignition.