Learn More
n-Pentane, Environmental Grade, 98+%
Suitable For Environmental, Pesticide, and GC Analysis | CAS: 109-66-0 | C5H12 | 72.15 g/mol
Supplier: Thermo Scientific Chemicals 040981K7
Description
n-Pentane is used in consumer merchandise such as spot lifters/cleaners and foaming shave gels and in commercial products such as a blowing agent for expanded polystyrene (foam) insulation. It may also be used as a solvent (in pure form) in laboratory, chemical analysis, and other settings where a highly volatile non-polar solvent is needed.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| n-Pentane | |
| Environmental Grade | |
| Colorless | |
| 0.626 g/cm3 at 20°C | |
| 4 L | |
| 260°C (500°F) | |
| 1.358 | |
| C5H12 | |
| 554 hPa (416mm Hg) at 20°C | |
| miscible with water. | |
| CCCCC | |
| 72.15 | |
| CHEBI:37830 | |
| Environmental |
| °C to 130°C | |
| 109-66-0 | |
| Colorless liquid | |
| 36°C | |
| -49°C (-57°F) | |
| Liquid | |
| >98% | |
| MFCD00009498 | |
| n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish | |
| OFBQJSOFQDEBGM-UHFFFAOYSA-N | |
| pentane | |
| 8003 | |
| 72.15 |
Chemical Identifiers
| 109-66-0 | |
| 72.15 | |
| OFBQJSOFQDEBGM-UHFFFAOYSA-N | |
| 8003 | |
| pentane |
| C5H12 | |
| MFCD00009498 | |
| n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish | |
| CHEBI:37830 | |
| CCCCC |
Safety and Handling
P210-P233-P235-P240-P241-P242-P243-P261-P271-P280-P301+P310-P303+P361+P353-P304+P340-P312-P331-P370+P378q-P501c
H225-H304-H336
DOTInformation : DOT Class: 3, Packaging Group: II
EINECSNumber : 203-692-4
RTECSNumber : RZ9450000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only