missing translation for 'onlineSavingsMsg'
Learn More

n-Pentane, 99+%, Extra Dry over Molecular Sieve, AcroSeal™

Catalog No. ac442990010
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 L
100 mL
Catalog No. Quantity
AC442990010 1 L
AC442991000 100 mL
2 options

Catalog No. AC442990010

Supplier: Thermo Scientific Chemicals 442990010

Only null left

CAS: 109-66-0 | C5H12 | 72.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 109-66-0
Molecular Formula C5H12
Molecular Weight (g/mol) 72.15
MDL Number MFCD00009498
InChI Key OFBQJSOFQDEBGM-UHFFFAOYSA-N
Synonym n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish
PubChem CID 8003
ChEBI CHEBI:37830
IUPAC Name pentane
SMILES CCCCC

Specifications

Melting Point -130.0°C
Density 0.6260g/mL
Boiling Point 36.0°C
Quantity 1 L
Flash Point −49°C
Refractive Index 1.358
Specific Gravity 0.626
Linear Formula CH3(CH2)3CH3
Viscosity 0.25 mPa.s (20°C)
Water 0.005% max.
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol and most other common organic, solvents
Formula Weight 72.15
Percent Purity ≥99%
Grade Extra Dry over Molecular Sieve
Chemical Name or Material n-Pentane
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement May be fatal if swallowed and enters airways. May cause drowsiness or dizziness. Toxic to aquatic life with long lasting effects. Highly flammable liquid and vapor. Repeated exposure may cause skin dryness or cracking.
Precautionary Statement GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician. Do NOT induce vomiting. Avoid release to the environment. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Ground/Bond container and receiving equipment.

RUO – Research Use Only