missing translation for 'onlineSavingsMsg'
Learn More

n-Pentane, 99+%, extra pure, anhydrous

Catalog No. p-2691963
Change view
Click to view available options
Quantity:
1 L
2.5 L
25 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC217240010 1 L
AC217240025 2.5 L
AC217240251 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC217240010 Supplier Thermo Scientific Chemicals Supplier No. 217240010
Only null left

CAS: 109-66-0 | C5H12 | 72.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 109-66-0
Molecular Formula C5H12
Molecular Weight (g/mol) 72.15
MDL Number MFCD00009498
InChI Key OFBQJSOFQDEBGM-UHFFFAOYSA-N
Synonym n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish
PubChem CID 8003
ChEBI CHEBI:37830
IUPAC Name pentane
SMILES CCCCC

Specifications

Melting Point -130.0°C
CAS Min % 99.0
CAS Max % 100.0
Density 0.6260g/mL
Quantity 1 L
Flash Point −49°C
Infrared Spectrum Authentic
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.358
Residue after Evaporation 0.001% max.
Specific Gravity 0.626
Assay Percent Range 99+%
Vapor Pressure Vapor Pressure: 573mbar at 20°C
Linear Formula CH3(CH2)3CH3
Viscosity 0.25 mPa.s (20°C)
Water 0.005% max.
Beilstein 01, 130
Merck Index 15, 7228
Boiling Point 36.0°C
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol and most other common organic, solvents
Formula Weight 72.15
Percent Purity ≥99%
Grade Extra Pure
Chemical Name or Material n-Pentane, p.a.
Show More Show Less
Hazard Category Danger
Hazard Statement GHS P Statement May be fatal if swallowed and enters airways. May cause drowsiness or dizziness. Toxic to aquatic life with long lasting effects. Highly flammable liquid and vapor. Repeated exposure may cause skin dryness or cracking.
Precautionary Statement GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician. Do NOT induce vomiting. Avoid release to the environment. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Ground/Bond container and receiving equipment.

RUO – Research Use Only