Learn More
n-Pentane, 99+%, Extra Dry, AcroSeal™
CAS: 109-66-0 | C5H12 | 72.15 g/mol
Supplier: Thermo Scientific Chemicals 397230010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| n-Pentane | |
| Extra Dry | |
| Colorless | |
| 100.0 | |
| 36.0°C | |
| −49°C | |
| AcroSeal™ Glass Bottle | |
| 1.358 | |
| 99+% | |
| C5H12 | |
| MFCD00009498 | |
| 0.002% max. | |
| n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish | |
| OFBQJSOFQDEBGM-UHFFFAOYSA-N | |
| pentane | |
| 8003 | |
| 72.15 | |
| Extra Dry |
| -130.0°C | |
| 109-66-0 | |
| 99.0 | |
| 0.6260g/mL | |
| 1 L | |
| Authentic | |
| Liquid | |
| 0.001% max. | |
| 0.626 | |
| CH3(CH2)3CH3 | |
| 0.25 mPa.s (20°C) | |
| 15, 7228 | |
| Solubility in water: insoluble. Other solubilities: soluble in alcohol and most other common organic, solvents | |
| CCCCC | |
| 72.15 | |
| CHEBI:37830 | |
| ≥99% |
Chemical Identifiers
| 109-66-0 | |
| 72.15 | |
| OFBQJSOFQDEBGM-UHFFFAOYSA-N | |
| 8003 | |
| pentane |
| C5H12 | |
| MFCD00009498 | |
| n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish | |
| CHEBI:37830 | |
| CCCCC |
Safety and Handling
GHS P Statement May be fatal if swallowed and enters airways. May cause drowsiness or dizziness. Toxic to aquatic life with long lasting effects. Highly flammable liquid and vapor. Repeated exposure may cause skin dryness or cracking.
GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician. Do NOT induce vomiting. Avoid release to the environment. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Ground/Bond container and receiving equipment.
Danger
RUO – Research Use Only