missing translation for 'onlineSavingsMsg'
Learn More

N-(p-Toluenesulfonyl)glycine, 97%

Catalog No. p-7035652
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5337106 5 g
AAH5337114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5337106 Supplier Thermo Scientific Chemicals Supplier No. H5337106

CAS: 1080-44-0 | C9H11NO4S | 229.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1080-44-0
Molecular Formula C9H11NO4S
Molecular Weight (g/mol) 229.25
MDL Number MFCD00045898
InChI Key VDKFCCZUCXYILI-UHFFFAOYSA-N
Synonym n-p-tosylglycine, tos-gly-oh, n-tosylglycine, 2-4-methylphenylsulfonamido acetic acid, n-4-methylphenyl sulfonyl glycine, n-p-toluenesulfonyl glycine, n-4-tosylglycine, glycine, n-4-methylphenyl sulfonyl, n-toluene-4-sulfonyl-glycine, toluene-4-sulfonylamino-acetic acid
PubChem CID 70653
IUPAC Name 2-[(4-methylphenyl)sulfonylamino]acetic acid
SMILES CC1=CC=C(C=C1)S(=O)(=O)NCC(=O)O

Specifications

Melting Point 146°C to 149°C
Quantity 5 g
Formula Weight 229.25
Percent Purity 97%
Chemical Name or Material N-(p-Toluenesulfonyl)glycine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only