missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Octyltrimethylammonium Chloride 98.0+%, TCI America™
Specifications
Specifications
| Color | White |
| Quantity | 25 g |
| Formula Weight | 207.79 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | n-Octyltrimethylammonium Chloride |
Safety and Handling
| EINECSNumber | (2)-0184&(9)-1971 |
| TSCA | No |