missing translation for 'onlineSavingsMsg'
Learn More

n-Octyltrimethylammonium Bromide 98.0+%, TCI America™

Catalog No. O016325G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O016325G 25 g
O0163500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O016325G Supplier TCI America Supplier No. O016325G

Chemical Identifiers

CAS 2083-68-3
Molecular Formula C11H26BrN
Molecular Weight (g/mol) 252.24
MDL Number MFCD00051873
InChI Key XCOHAFVJQZPUKF-UHFFFAOYSA-M
Synonym n-octyltrimethylammonium bromide, octalone, octyltrimethylammonium bromide, n,n,n-trimethyloctan-1-aminium bromide, 1-octanaminium, n,n,n-trimethyl-, bromide, n,n,n-trimethyl-1-octanaminium bromide, trimethyloctylammonium bromide 6ci,7ci, n-octyltrimethylammoniumbromide, trimethyloctylamine, bromide, trimethyl octyl azanium bromide
PubChem CID 74964
ChEBI CHEBI:346954
IUPAC Name trimethyl(octyl)azanium;bromide
SMILES CCCCCCCC[N+](C)(C)C.[Br-]

Specifications

Melting Point 217°C
Color White
Quantity 25 g
Formula Weight 252.24
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material n-Octyltrimethylammonium Bromide
EINECSNumber (2)-0184&(9)-1971
RTECSNumber RG8275000
TSCA No