Learn More
n-Octyltrimethoxysilane, 97+%
CAS: 3069-40-7 | C11H26O3Si | 234.41 g/mol
Supplier: Thermo Scientific Chemicals 04269814
Description
n-Octyltrimethoxysilane is used in medicine field, electronic industry and polymer material. It is also used as surface modifier.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 75°C (0.1mmHg) | |
| 3069-40-7 | |
| Liquid | |
| C11H26O3Si | |
| 102°C (216°F) | |
| Soluble in common non-polar organic solvent. | |
| CCCCCCCC[Si](OC)(OC)OC | |
| 234.41 | |
| 234.41 | |
| 1.417 | |
| 0.907 g/mL |
| n-Octyltrimethoxysilane | |
| Colorless to Yellow | |
| 25 g | |
| MFCD00043053 | |
| trimethoxy octyl silane, n-octyltrimethoxysilane, octyltrimethoxysilane, silane, trimethoxyoctyl, unii-fz07e4lw2m, octyl-trimethoxysilane, fz07e4lw2m, trimethoxycaprylylsilane, trimethoxy-octylsilane, trimethoxy-n-octylsilane | |
| NMEPHPOFYLLFTK-UHFFFAOYSA-N | |
| trimethoxy(octyl)silane | |
| 76485 | |
| ≥97% | |
| Moisture sensitive |
Chemical Identifiers
| 3069-40-7 | |
| 234.41 | |
| NMEPHPOFYLLFTK-UHFFFAOYSA-N | |
| 76485 | |
| CCCCCCCC[Si](OC)(OC)OC |
| C11H26O3Si | |
| MFCD00043053 | |
| trimethoxy octyl silane, n-octyltrimethoxysilane, octyltrimethoxysilane, silane, trimethoxyoctyl, unii-fz07e4lw2m, octyl-trimethoxysilane, fz07e4lw2m, trimethoxycaprylylsilane, trimethoxy-octylsilane, trimethoxy-n-octylsilane | |
| trimethoxy(octyl)silane |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P311-P332+P313-P362-P501c
H315-H319-H331
EINECSNumber : 221-338-7
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only