missing translation for 'onlineSavingsMsg'
Learn More

n-Octyltriethoxysilane, 95%

Catalog No. AAL0440718
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0440718 50 g
AAL0440730 250 g
AAL044070B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0440718 Supplier Thermo Scientific Chemicals Supplier No. L0440718

CAS: 2943-75-1 | C14H32O3Si | 276.49 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2943-75-1
Molecular Formula C14H32O3Si
Molecular Weight (g/mol) 276.49
MDL Number MFCD00039883
InChI Key MSRJTTSHWYDFIU-UHFFFAOYSA-N
Synonym n-octyltriethoxysilane, octyltriethoxysilane, triethoxy octyl silane, silane, triethoxyoctyl, dynasylan octeo, octyl triethoxy silane, prosil 9202, silquest a 137, prosil 9234, a 137 coupling agent
PubChem CID 76262
IUPAC Name triethoxy(octyl)silane
SMILES CCCCCCCC[Si](OCC)(OCC)OCC

Specifications

Boiling Point 265°C
Melting Point <-40°C
Quantity 50 g
Beilstein 2325287
Flash Point 109°C (228°F)
Formula Weight 276.49
Percent Purity 95%
Refractive Index 1.417
Sensitivity Moisture sensitive
Density 0.878
Chemical Name or Material n-Octyltriethoxysilane
Show More Show Less
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 220-941-2
RTECSNumber VV6695500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only