missing translation for 'onlineSavingsMsg'
Learn More

n-Octylsuccinic Anhydride 98.0+%, TCI America™

Catalog No. O004925G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O004925G 25 g
O0049250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O004925G Supplier TCI America Supplier No. O004925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4200-92-4
Molecular Formula C12H20O3
Molecular Weight (g/mol) 212.29
MDL Number MFCD00014553
InChI Key OAJCSERLBQROJC-UHFFFAOYNA-N
PubChem CID 77869
IUPAC Name 3-octyloxolane-2,5-dione
SMILES CCCCCCCCC1CC(=O)OC1=O

Specifications

Melting Point 65°C
Color White-Yellow
Quantity 25 g
Physical Form Crystal-Powder at 20°C
Formula Weight 212.29
Percent Purity ≥98.0% (GC,T)
Chemical Name or Material n-Octylsuccinic Anhydride