missing translation for 'onlineSavingsMsg'
Learn More

n-Octylphosphonic Acid 98.0+%, TCI America™

Catalog No. O03805G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O03805G 5 g
O03801G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O03805G Supplier TCI America Supplier No. O03805G

Chemical Identifiers

CAS 4724-48-5
Molecular Formula C8H19O3P
Molecular Weight (g/mol) 194.211
MDL Number MFCD00015841
InChI Key NJGCRMAPOWGWMW-UHFFFAOYSA-N
Synonym n-octylphosphonic acid, 1-octylphosphonic acid, phosphonic acid, octyl, phosphonic acid, p-octyl, unii-95c2aw87bo, octylphosphonicacid, acmc-209uni, p-octyl-phosphonic acid, octylphosphonic acid, dsstox_cid_24679
PubChem CID 78452
IUPAC Name octylphosphonic acid
SMILES CCCCCCCCP(=O)(O)O

Specifications

Melting Point 101°C
Color White-Yellow
Quantity 5 g
UN Number 3261
Formula Weight 194.21
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material n-Octylphosphonic Acid
EINECSNumber (2)-1995
TSCA Yes