missing translation for 'onlineSavingsMsg'
Learn More

n-Octyl p-Toluenesulfonate 96.0+%, TCI America™

Catalog No. T088525G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T088525G 25 g
T0885500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T088525G Supplier TCI America Supplier No. T088525G

Chemical Identifiers

CAS 3386-35-4
Molecular Formula C15H24O3S
Molecular Weight (g/mol) 284.414
MDL Number MFCD00059447
InChI Key LYQJBZLAANNIER-UHFFFAOYSA-N
Synonym p-Toluenesulfonic Acid n-Octyl Ester
PubChem CID 599296
IUPAC Name octyl 4-methylbenzenesulfonate
SMILES CCCCCCCCOS(=O)(=O)C1=CC=C(C=C1)C

Specifications

Color Yellow
Quantity 25 g
Formula Weight 284.41
Percent Purity ≥96.0% (GC,T)
Physical Form Liquid
Chemical Name or Material n-Octyl p-Toluenesulfonate