Learn More
N-Octyl-N,N-dimethyl-3-ammonio-1-propanesulfonate, 98%, Thermo Scientific™
$152.65 - $484.18
Chemical Identifiers
| CAS | 15178-76-4 |
|---|---|
| Molecular Formula | C13H29NO3S |
| Molecular Weight (g/mol) | 279.44 |
| InChI Key | QZRAABPTWGFNIU-UHFFFAOYSA-N |
| Synonym | octyl sulfobetaine, 3-dimethyl-octylazaniumyl propane-1-sulfonate, n-octyl-n,n-dimethyl-3-ammonio-1-propanesulfonate, 3-n,n-dimethyloctylammonio propanesulfonate inner salt, 3-dimethyl octyl ammonio propane-1-sulfonate, 3-n,n-dimethyloctylammonio propanesulfonate solution, n-octyl-n,n-dimethyl-3-ammonio-1-propanesulfonate sb-8, n-octylsulfobetaine, zwittergent 3-08 |
| PubChem CID | 3084219 |
| IUPAC Name | 3-[dimethyl(octyl)azaniumyl]propane-1-sulfonate |
| SMILES | CCCCCCCC[N+](C)(C)CCCS(=O)(=O)[O-] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AC459700050
|
N/A | 5g |
Each for $152.65
|
|
|||||
|
AC459700250
|
N/A | 25g |
Each for $484.18
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 15178-76-4 | |
| 279.44 | |
| octyl sulfobetaine, 3-dimethyl-octylazaniumyl propane-1-sulfonate, n-octyl-n,n-dimethyl-3-ammonio-1-propanesulfonate, 3-n,n-dimethyloctylammonio propanesulfonate inner salt, 3-dimethyl octyl ammonio propane-1-sulfonate, 3-n,n-dimethyloctylammonio propanesulfonate solution, n-octyl-n,n-dimethyl-3-ammonio-1-propanesulfonate sb-8, n-octylsulfobetaine, zwittergent 3-08 | |
| 3-[dimethyl(octyl)azaniumyl]propane-1-sulfonate |
| C13H29NO3S | |
| QZRAABPTWGFNIU-UHFFFAOYSA-N | |
| 3084219 | |
| CCCCCCCC[N+](C)(C)CCCS(=O)(=O)[O-] |
Specifications
| 15178-76-4 | |
| Authentic | |
| C13H29NO3S | |
| (10% in water) Clear solution | |
| CCCCCCCC[N+](C)(C)CCCS(=O)(=O)[O-] | |
| 279.44 | |
| 279.44 | |
| N-Octyl-N,N-dimethyl-3-ammonio-1-propanesulfonate |
| 2% max. | |
| 97.5% min. (HPLC) | |
| octyl sulfobetaine, 3-dimethyl-octylazaniumyl propane-1-sulfonate, n-octyl-n,n-dimethyl-3-ammonio-1-propanesulfonate, 3-n,n-dimethyloctylammonio propanesulfonate inner salt, 3-dimethyl octyl ammonio propane-1-sulfonate, 3-n,n-dimethyloctylammonio propanesulfonate solution, n-octyl-n,n-dimethyl-3-ammonio-1-propanesulfonate sb-8, n-octylsulfobetaine, zwittergent 3-08 | |
| QZRAABPTWGFNIU-UHFFFAOYSA-N | |
| 3-[dimethyl(octyl)azaniumyl]propane-1-sulfonate | |
| 3084219 | |
| 98% |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee
GHS Signal Word: Warning
RUO – Research Use Only