missing translation for 'onlineSavingsMsg'
Learn More

n-Octyl acetate, 98+%

Catalog No. AAL0640518
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0640518 50 g
AAL0640530 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0640518 Supplier Thermo Scientific Chemicals Supplier No. L0640518
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 112-14-1 | C10H20O2 | 172.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 112-14-1
Molecular Formula C10H20O2
Molecular Weight (g/mol) 172.27
MDL Number MFCD00009562
InChI Key YLYBTZIQSIBWLI-UHFFFAOYSA-N
Synonym n-octyl acetate, acetic acid, octyl ester, caprylyl acetate, 1-octyl acetate, acetic acid octyl ester, octyl ethanoate, n-octanyl acetate, n-octyl ethanoate, 1-octanol acetate, acetate c-8
PubChem CID 8164
ChEBI CHEBI:87495
IUPAC Name octyl acetate
SMILES CCCCCCCCOC(C)=O

Specifications

Density 0.868
Boiling Point 210°C to 211°C
Flash Point 86°C (186°F)
Refractive Index 1.419
Linear Formula CH3CO2(CH2)7CH3
Quantity 50 g
Beilstein 1754554
Formula Weight 172.27
Percent Purity ≥98%
Chemical Name or Material n-Octyl acetate
Hazard Category H227-H315-H319
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P370+P378q-P501c
EINECSNumber 203-939-6
RTECSNumber AJ1400000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only