missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Octyl 4,6-Dibromo-3-fluorothieno[3,4-b]thiophene-2-carboxylate 94.0+%, TCI America™
Specifications
Specifications
| Melting Point | 55°C |
| Quantity | 100 mg |
| Formula Weight | 472.23 |
| Percent Purity | ≥94.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | n-Octyl 4,6-Dibromo-3-fluorothieno[3,4-b]thiophene-2-carboxylate |
Safety and Handling
| TSCA | No |