missing translation for 'onlineSavingsMsg'
Learn More

n-Octanohydrazide 97.0+%, TCI America™

Catalog No. O03245G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O03245G 5 g
O032425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O03245G Supplier TCI America Supplier No. O03245G
Only null left

Chemical Identifiers

CAS 6304-39-8
Molecular Formula C8H18N2O
Molecular Weight (g/mol) 158.25
MDL Number MFCD00011588
InChI Key VMUZVGRNTPFTKE-UHFFFAOYSA-N
Synonym n-Octanoic Hydrazide
PubChem CID 80556
IUPAC Name octanehydrazide
SMILES CCCCCCCC(=O)NN

Specifications

Melting Point 90°C
Color White-Yellow
Quantity 5 g
Formula Weight 158.25
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material n-Octanohydrazide