missing translation for 'onlineSavingsMsg'
Learn More

n-Octanal 98.0+%, TCI America™

Catalog No. p-7134151
Change view
Click to view available options
Quantity:
25 mL
500 mL
Catalog No. Quantity
O004425ML 25 mL
O0044500ML 500 mL
2 options

Catalog No. O004425ML

Supplier: TCI America O004425ML

Only null left

Chemical Identifiers

CAS 124-13-0
Molecular Formula C8H16O
Molecular Weight (g/mol) 128.22
MDL Number MFCD00007029
InChI Key NUJGJRNETVAIRJ-UHFFFAOYSA-N
Synonym caprylic aldehyde, caprylaldehyde, n-octanal, 1-octanal, n-octyl aldehyde, n-octaldehyde, n-caprylaldehyde, octanaldehyde, aldehyde c-8, n-octylal
PubChem CID 454
ChEBI CHEBI:17935
IUPAC Name octanal
SMILES CCCCCCCC=O

Specifications

Color Yellow
Boiling Point 170°C
Quantity 25 mL
UN Number 1191
Formula Weight 128.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material n-Octanal
EINECSNumber (2)-0494
RTECSNumber RG7780000
TSCA Yes