missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Octacosane, 99%
CAS: 630-02-4 | C28H58 | 394.77 g/mol
Supplier: Thermo Scientific Chemicals 04246018
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| n-Octacosane | |
| 61°C to 63°C | |
| 227°C (441°F) | |
| MFCD00009355 | |
| n-octacosane, n-octcosane, unii-vff49836p8, ch3-ch2 26-ch3, octacosane, analytical standard, ccris 680, octacosane, n, octacosane, heptacosane, methyl, n-octacosane 10g | |
| ZYURHZPYMFLWSH-UHFFFAOYSA-N | |
| octacosane | |
| 12408 | |
| 394.77 | |
| Crystalline |
| 630-02-4 | |
| 440°C | |
| C28H58 | |
| 50 g | |
| Soluble in chloroform,acetone,benzene,and toluene. Insoluble in water. | |
| CCCCCCCCCCCCCCCCCCCCCCCCCCCC | |
| 394.77 | |
| CHEBI:32943 | |
| 99% |
Chemical Identifiers
| 630-02-4 | |
| 394.77 | |
| ZYURHZPYMFLWSH-UHFFFAOYSA-N | |
| 12408 | |
| octacosane |
| C28H58 | |
| MFCD00009355 | |
| n-octacosane, n-octcosane, unii-vff49836p8, ch3-ch2 26-ch3, octacosane, analytical standard, ccris 680, octacosane, n, octacosane, heptacosane, methyl, n-octacosane 10g | |
| CHEBI:32943 | |
| CCCCCCCCCCCCCCCCCCCCCCCCCCCC |
Safety and Handling
EINECSNumber : 211-125-7
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only