missing translation for 'onlineSavingsMsg'
Learn More

n-Octacosane, 99%

Catalog No. AA4246030
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4246030 250 g
AA4246009 10 g
AA4246018 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4246030 Supplier Thermo Scientific Chemicals Supplier No. 04246030

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 630-02-4 | C28H58 | 394.77 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 630-02-4
Molecular Formula C28H58
Molecular Weight (g/mol) 394.77
MDL Number MFCD00009355
InChI Key ZYURHZPYMFLWSH-UHFFFAOYSA-N
Synonym n-octacosane, n-octcosane, unii-vff49836p8, ch3-ch2 26-ch3, octacosane, analytical standard, ccris 680, octacosane, n, octacosane, heptacosane, methyl, n-octacosane 10g
PubChem CID 12408
ChEBI CHEBI:32943
IUPAC Name octacosane
SMILES CCCCCCCCCCCCCCCCCCCCCCCCCCCC

Specifications

Melting Point 61°C to 63°C
Boiling Point 440°C
Flash Point 227°C (441°F)
Quantity 250 g
Solubility Information Soluble in chloroform,acetone,benzene,and toluene. Insoluble in water.
Formula Weight 394.77
Percent Purity 99%
Physical Form Crystalline
Chemical Name or Material n-Octacosane
EINECSNumber 211-125-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only