missing translation for 'onlineSavingsMsg'
Learn More

N,O-Dimethylhydroxylamine Hydrochloride 98.0+%, TCI America™

Catalog No. p-7126999
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D189925G 25 g
D1899100G 100 g
D1899500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D189925G Supplier TCI America Supplier No. D189925G
Only null left

Chemical Identifiers

CAS 6638-79-5
Molecular Formula C2H8ClNO
Molecular Weight (g/mol) 97.54
MDL Number MFCD00012485
InChI Key USZLCYNVCCDPLQ-UHFFFAOYSA-N
Synonym n,o-dimethylhydroxylamine hydrochloride, n,o-dimethylhydroxylamine hcl, n-methoxymethylamine hydrochloride, methanamine, n-methoxy-, hydrochloride, o,n-dimethylhydroxylamine hydrochloride, n-methoxymethanamine hydrochloride, o,n-dimethyl-hydroxylamine hydrochloride, methoxy methyl amine hydrochloride, n,o-dimethylhydroxyamine hydrochloride, n,o-dimethylhydroxylaminehydrochloride
PubChem CID 81138
IUPAC Name hydrogen methoxy(methyl)amine chloride
SMILES [H+].[Cl-].CNOC

Specifications

Melting Point 115°C
Color White
Quantity 25 g
Formula Weight 97.54
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N,O-Dimethylhydroxylamine Hydrochloride