missing translation for 'onlineSavingsMsg'
Learn More

N,O-Dimethylhydroxylamine hydrochloride, 98%

Catalog No. p-118372
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC116340050 5 g Glass bottle
AC116340250 25 g Glass bottle
AC116341000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC116340050 Supplier Thermo Scientific Chemicals Supplier No. 116340050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6638-79-5 | C2H8ClNO | 97.54 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent

Chemical Identifiers

CAS 6638-79-5
Molecular Formula C2H8ClNO
Molecular Weight (g/mol) 97.54
MDL Number MFCD00012485
InChI Key USZLCYNVCCDPLQ-UHFFFAOYSA-N
Synonym n,o-dimethylhydroxylamine hydrochloride, n,o-dimethylhydroxylamine hcl, n-methoxymethylamine hydrochloride, methanamine, n-methoxy-, hydrochloride, o,n-dimethylhydroxylamine hydrochloride, n-methoxymethanamine hydrochloride, o,n-dimethyl-hydroxylamine hydrochloride, methoxy methyl amine hydrochloride, n,o-dimethylhydroxyamine hydrochloride, n,o-dimethylhydroxylaminehydrochloride
PubChem CID 81138
SMILES [H+].[Cl-].CNOC

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 112.0°C to 116.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula CH3ONHCH3·HCl
Quantity 5 g
Beilstein 04, 534
Fieser 11,201; 12,205; 15,144
Solubility Information (10% in water) Clear
IUPAC Name N-methoxymethanamine;hydrochloride
Formula Weight 97.55
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material N, O-Dimethylhydroxylamine hydrochloride, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Wear protective gloves/protective clothing/eye protec
EINECSNumber 229-642-1
TSCA TSCA

RUO – Research Use Only