missing translation for 'onlineSavingsMsg'
Learn More

N,O-Bis(trimethylsilyl)trifluoroacetamide 95.0+%, TCI America™

Supplier:  TCI America B08305ML

 View more versions of this product

Catalog No. B08305ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

N,O-Bis(trimethylsilyl)trifluoroacetamide
Yellow
C8H18F3NOSi2
5 mL
bstfa, n,o-bis trimethylsilyl trifluoroacetamide, acetamide, 2,2,2-trifluoro-o,n-bis trimethylsilyl, trimethylsilyl 2,2,2-trifluoro-n-trimethylsilyl acetimidate, trimethylsilyl 2,2,2-trifluoro-n-trimethylsilyl ethanimidate, trimethylsilyl 1z-2,2,2-trifluoro-n-z-trimethylsilyl ethanimidoate #, n,o-bis trimethylsilyl trifluoroacetamide bstfa, trimethylsilyl 2,2,2-trifluoro-n-trimethylsilyl ethanecarboximidate
C[Si](C)(C)N=C(C(F)(F)F)O[Si](C)(C)C
257.403
257.40
Liquid
25561-30-2
142°C
MFCD00008269
2924
XCOBLONWWXQEBS-GHXNOFRVSA-N
trimethylsilyl (1Z)-2,2,2-trifluoro-N-trimethylsilylethanimidate
9601896
≥95.0% (GC)

Chemical Identifiers

25561-30-2
257.403
XCOBLONWWXQEBS-GHXNOFRVSA-N
9601896
C[Si](C)(C)N=C(C(F)(F)F)O[Si](C)(C)C
C8H18F3NOSi2
MFCD00008269
bstfa, n,o-bis trimethylsilyl trifluoroacetamide, acetamide, 2,2,2-trifluoro-o,n-bis trimethylsilyl, trimethylsilyl 2,2,2-trifluoro-n-trimethylsilyl acetimidate, trimethylsilyl 2,2,2-trifluoro-n-trimethylsilyl ethanimidate, trimethylsilyl 1z-2,2,2-trifluoro-n-z-trimethylsilyl ethanimidoate #, n,o-bis trimethylsilyl trifluoroacetamide bstfa, trimethylsilyl 2,2,2-trifluoro-n-trimethylsilyl ethanecarboximidate
trimethylsilyl (1Z)-2,2,2-trifluoro-N-trimethylsilylethanimidate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-3613

TSCA : No

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions