missing translation for 'onlineSavingsMsg'
Learn More

N,O-Bis(trimethylsilyl)trifluoroacetamide, 99+%, packaged under Argon in resealable ChemSeal™ bottles

Catalog No. p-7052035
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4242906 5 g
AA4242914 25 g
AA4242922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4242906 Supplier Thermo Scientific Chemicals Supplier No. 04242906
Only null left

CAS: 25561-30-2 | C8H18F3NOSi2 | 257.403 g/mol

N,O-Bis(trimethylsilyl)trifluoroacetamide is used as a derivatization reagent for gas chromatography and mass-spectrometry analysis of phenolic acids in fruits. It is also used as a reactant in the preparation of pyrimidinone ribosides and analogs which shows anti-tumor properties. It plays as an important role as a reagent for the regioselective desulfation of polysaccharide sulfates. Furthermore, it is used to prepare trimethylsilyl ethers of carbohydrates and alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon

Chemical Identifiers

CAS 25561-30-2
Molecular Formula C8H18F3NOSi2
Molecular Weight (g/mol) 257.403
MDL Number MFCD00008269
InChI Key XCOBLONWWXQEBS-GHXNOFRVSA-N
Synonym bstfa, n,o-bis trimethylsilyl trifluoroacetamide, acetamide, 2,2,2-trifluoro-o,n-bis trimethylsilyl, trimethylsilyl 2,2,2-trifluoro-n-trimethylsilyl acetimidate, trimethylsilyl 2,2,2-trifluoro-n-trimethylsilyl ethanimidate, trimethylsilyl 1z-2,2,2-trifluoro-n-z-trimethylsilyl ethanimidoate #, n,o-bis trimethylsilyl trifluoroacetamide bstfa, trimethylsilyl 2,2,2-trifluoro-n-trimethylsilyl ethanecarboximidate
PubChem CID 9601896
IUPAC Name trimethylsilyl (1Z)-2,2,2-trifluoro-N-trimethylsilylethanimidate
SMILES C[Si](C)(C)N=C(C(F)(F)F)O[Si](C)(C)C

Specifications

Melting Point -10°C
Color Colorless to Yellow
Density 0.97 g/mL
Boiling Point 145°C to 147°C
Flash Point 23°C (73°F)
Packaging ChemSeal™ bottles
Refractive Index 1.384
Quantity 5 g
UN Number UN2924
Beilstein 3607620
Sensitivity Moisture sensitive
Solubility Information Miscible with chloroform.
Formula Weight 257.4
Percent Purity ≥99%
Physical Form Liquid
Chemical Name or Material N,O-Bis (trimethylsilyl)trifluoroacetamide
Show More Show Less
Hazard Category H226-H314-H335
Hazard Statement GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Hazard Class: 3
EINECSNumber 247-103-9
TSCA Yes
storageNote1 2°C to 8°C
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only