missing translation for 'onlineSavingsMsg'
Learn More

N,O-Bis(trimethylsilyl)trifluoroacetamide, with 1% TMCS, packaged under Argon in resealable ChemSeal™ bottles

Catalog No. AA4392806
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4392806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA4392806 Supplier Thermo Scientific Chemicals Supplier No. 04392806

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 25561-30-2 | C8H18F3NOSi2 | 257.403 g/mol

N,O-Bis(trimethylsilyl)trifluoroacetamide is used for analytical purposes or as a chemical reagent for synthesis of more complex molecules. It is silylation reagent useful in GC and GC-MS for the derivatization of a wide range of functional groups under mild conditions. It is also used as DAT inhibitor and a SLC6A2 inhibitor. It is routinely used to derivatize carboxylic acids, phenols, steroids, amines, alcohols, and alkaloids and amides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon

Chemical Identifiers

CAS 25561-30-2
Molecular Formula C8H18F3NOSi2
Molecular Weight (g/mol) 257.403
MDL Number MFCD00008269
InChI Key XCOBLONWWXQEBS-GHXNOFRVSA-N
Synonym bstfa, n,o-bis trimethylsilyl trifluoroacetamide, acetamide, 2,2,2-trifluoro-o,n-bis trimethylsilyl, trimethylsilyl 2,2,2-trifluoro-n-trimethylsilyl acetimidate, trimethylsilyl 2,2,2-trifluoro-n-trimethylsilyl ethanimidate, trimethylsilyl 1z-2,2,2-trifluoro-n-z-trimethylsilyl ethanimidoate #, n,o-bis trimethylsilyl trifluoroacetamide bstfa, trimethylsilyl 2,2,2-trifluoro-n-trimethylsilyl ethanecarboximidate
PubChem CID 9601896
IUPAC Name trimethylsilyl (1Z)-2,2,2-trifluoro-N-trimethylsilylethanimidate
SMILES C[Si](C)(C)N=C(C(F)(F)F)O[Si](C)(C)C

Specifications

Melting Point -10°C
Color Colorless to Yellow
Density 0.97 g/mL
Boiling Point 145°C to 147°C
Flash Point 23°C (73°F)
Packaging ChemSeal™ bottles
Refractive Index 1.384
Quantity 5 g
UN Number UN2924
Beilstein 3607620
Sensitivity Moisture sensitive
Solubility Information Soluble in water (hydrolyses),chloroform and hexane.
Formula Weight 257.4
Physical Form Liquid
Chemical Name or Material N,O-Bis (trimethylsilyl)trifluoroacetamide, With 1% TMCS
Show More Show Less
Hazard Category H226-H314
Hazard Statement GHS H Statement
H314-H226
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Precautionary Statement P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
EINECSNumber 247-103-9
TSCA Yes
storageNote1
  • Packaged under Argon
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only