missing translation for 'onlineSavingsMsg'
Learn More

N,O-Bis(trimethylsilyl)acetamide 80.0+%, TCI America™

Catalog No. p-7118489
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
10 mL
100 mL
Catalog No. Quantity
B051110ML 10 mL
B0511100ML 100 mL
2 options

Catalog No. B051110ML

Supplier: TCI America B051110ML

Only null left

Chemical Identifiers

CAS 10416-59-8
Molecular Formula C8H21NOSi2
Molecular Weight (g/mol) 203.43
MDL Number MFCD00008270
InChI Key SIOVKLKJSOKLIF-UHFFFAOYSA-N
Synonym n,o-bis trimethylsilyl acetamide, trimethylsilyl 1z-n-trimethylsilylethanimidate
PubChem CID 6913588
IUPAC Name trimethylsilyl N-(trimethylsilyl)ethanimidate
SMILES CC(O[Si](C)(C)C)=N[Si](C)(C)C

Specifications

Melting Point -24°C
Color Yellow
Boiling Point 73°C
Quantity 10 mL
UN Number 2920
Formula Weight 203.43
Percent Purity ≥80.0% (GC)
Physical Form Liquid
Chemical Name or Material N,O-Bis(trimethylsilyl)acetamide
EINECSNumber (2)-3273
RTECSNumber AK3000000
TSCA Yes