missing translation for 'onlineSavingsMsg'
Learn More

N,O-Bis(trimethylsilyl)acetamide Kit BSA 1 mL * 8 / Reaction vial, capacity 2 mL * 8 80.0+%, TCI America™

Catalog No. B0911KIT
Change view
Click to view available options
Quantity:
Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0911KIT Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B0911KIT Supplier TCI America Supplier No. B09111KIT

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10416-59-8
Molecular Formula C8H21NOSi2
Molecular Weight (g/mol) 203.43
MDL Number MFCD00008270
InChI Key SIOVKLKJSOKLIF-UHFFFAOYSA-N
Synonym n,o-bis trimethylsilyl acetamide, trimethylsilyl 1z-n-trimethylsilylethanimidate
PubChem CID 6913588
IUPAC Name trimethylsilyl N-(trimethylsilyl)ethanimidate
SMILES CC(O[Si](C)(C)C)=N[Si](C)(C)C

Specifications

Melting Point -24°C
Color Yellow
Boiling Point 73°C
Quantity Kit
UN Number 2920
Percent Purity ≥80.0% (GC)
Physical Form Liquid
Chemical Name or Material N,O-Bis(trimethylsilyl)acetamide Kit BSA 1 mL * 8 / Reaction vial, capacity 2 mL * 8
EINECSNumber (2)-3273
RTECSNumber AK3000000
TSCA Yes