missing translation for 'onlineSavingsMsg'
Learn More

N,O-Bis(trimethylsilyl)acetamide, 95%

Catalog No. AAL0018314
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0018314 25 g
AAL0018322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0018314 Supplier Thermo Scientific Chemicals Supplier No. L0018314
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 10416-59-8 | C8H21NOSi2 | 203.43 g/mol

N,O-Bis(trimethylsilyl)acetamide is used as a regioselective desulfation reagent as well as a preparatory agent for carbohydrate and alcohol trimethylsilyl ethers. It is also used for the derivatization of polar functional groups such as carboxylic acids, phenols, steroids, amines, alcohols, alkaloids and amides. It is employed in the formation of stable trimethylsilyl derivatives. Further, it is used in analytical chemistry for the derivatization of compounds in analysis to increase their volatility and to introduce the trimethylsilyl protecting group in organic synthesis. In addition to this, it is used in the protection of hydroxyl groups in natural products, functional groups in organic intermediates and derivatization reagent for gas chromatography-Mass spectral analysis of phenolic acids in fruits.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,O-Bis(trimethylsilyl)acetamide is used as a regioselective desulfation reagent as well as a preparatory agent for carbohydrate and alcohol trimethylsilyl ethers. It is also used for the derivatization of polar functional groups such as carboxylic acids, phenols, steroids, amines, alcohols, alkaloids and amides. It is employed in the formation of stable trimethylsilyl derivatives. Further, it is used in analytical chemistry for the derivatization of compounds in analysis to increase their volatility and to introduce the trimethylsilyl protecting group in organic synthesis. In addition to this, it is used in the protection of hydroxyl groups in natural products, functional groups in organic intermediates and derivatization reagent for gas chromatography-Mass spectral analysis of phenolic acids in fruits.

Solubility
Miscible with many nonpolar and polar aprotic solvents.

Notes
Air and moisture sensitive. Incompatible with strong oxidizing agents, water and acids.

Chemical Identifiers

CAS 10416-59-8
Molecular Formula C8H21NOSi2
Molecular Weight (g/mol) 203.43
MDL Number MFCD00008270
InChI Key SIOVKLKJSOKLIF-UHFFFAOYSA-N
Synonym n,o-bis trimethylsilyl acetamide, trimethylsilyl 1z-n-trimethylsilylethanimidate
PubChem CID 6913588
SMILES CC(O[Si](C)(C)C)=N[Si](C)(C)C

Specifications

Boiling Point 71°C to 73°C (35mmHg)
Melting Point -24°C
Quantity 25 g
UN Number UN2920
Beilstein 1306669
Flash Point 42°C (107°F)
Solubility Information Miscible with many nonpolar and polar aprotic solvents.
IUPAC Name trimethylsilyl N-(trimethylsilyl)ethanimidate
Formula Weight 203.43
Percent Purity 95%
Refractive Index 1.417
Sensitivity Moisture sensitive
Density 0.829
Chemical Name or Material N,O-Bis(trimethylsilyl)acetamide
Show More Show Less
Hazard Category H226-H302-H314-H335
Hazard Statement GHS H Statement
H301-H314-H318-H226
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, FLAMMABLE, N.O.S.
EINECSNumber 233-892-7
RTECSNumber AK3000000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only