missing translation for 'onlineSavingsMsg'
Learn More

N,O-Bis(trimethylsilyl)acetamide (25% in Acetonitrile), TCI America™

Catalog No. B051012ML
Change view
Click to view available options
Quantity:
12 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B051012ML 12 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B051012ML Supplier TCI America Supplier No. B051012ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10416-59-8
Molecular Formula C8H21NOSi2
Molecular Weight (g/mol) 203.43
MDL Number MFCD00008270
InChI Key SIOVKLKJSOKLIF-UHFFFAOYSA-N
Synonym n,o-bis trimethylsilyl acetamide, trimethylsilyl 1z-n-trimethylsilylethanimidate
PubChem CID 6913588
IUPAC Name trimethylsilyl N-(trimethylsilyl)ethanimidate
SMILES CC(O[Si](C)(C)C)=N[Si](C)(C)C

Specifications

Color Colorless
Quantity 12 mL
UN Number 1992
Physical Form Liquid
Chemical Name or Material N,O-Bis(trimethylsilyl)acetamide (25% in Acetonitrile) [Trimethylsilylating Reagent, for NH2 compoun
EINECSNumber (2)-3273
RTECSNumber AK3000000
TSCA Yes
Recommended Storage Refrigerator