missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,O-Bis(tert-butoxycarbonyl)hydroxylamine 98.0+%, TCI America™
$50.33 - $141.67
Chemical Identifiers
| CAS | 85006-25-3 |
|---|---|
| Molecular Formula | C10H19NO5 |
| Molecular Weight (g/mol) | 233.26 |
| MDL Number | MFCD00034797 |
| InChI Key | AGOSGCWATIJZHQ-UHFFFAOYSA-N |
| Synonym | tert-butyl tert-butoxycarbonyl oxycarbamate, n,o-di-boc-hydroxylamine, tert-butyl n-tert-butoxycarbonyloxy carbamate, n,o-bis tert-butoxycarbonyl hydroxylamine, tert-butyl tert-butoxycarbonyl oxy carbamate, tert-butyl 1,1-dimethylethoxy carbonyl oxy carbamate, tert-butyl tert-butoxycarbonyloxy carbamate, tert-butoxycarbonylamino tert-butyl carbonate, tert-butyl 2-methylpropan-2-yl oxycarbonylamino carbonate, 2-tert-butoxy carbonyl amino oxy carbonyl oxy-2-methylpropane |
| PubChem CID | 2728863 |
| IUPAC Name | 2-{[({[(tert-butoxy)carbonyl]amino}oxy)carbonyl]oxy}-2-methylpropane |
| SMILES | CC(C)(C)OC(=O)NOC(=O)OC(C)(C)C |
Chemical Identifiers
| 85006-25-3 | |
| 233.26 | |
| AGOSGCWATIJZHQ-UHFFFAOYSA-N | |
| 2728863 | |
| CC(C)(C)OC(=O)NOC(=O)OC(C)(C)C |
| C10H19NO5 | |
| MFCD00034797 | |
| tert-butyl tert-butoxycarbonyl oxycarbamate, n,o-di-boc-hydroxylamine, tert-butyl n-tert-butoxycarbonyloxy carbamate, n,o-bis tert-butoxycarbonyl hydroxylamine, tert-butyl tert-butoxycarbonyl oxy carbamate, tert-butyl 1,1-dimethylethoxy carbonyl oxy carbamate, tert-butyl tert-butoxycarbonyloxy carbamate, tert-butoxycarbonylamino tert-butyl carbonate, tert-butyl 2-methylpropan-2-yl oxycarbonylamino carbonate, 2-tert-butoxy carbonyl amino oxy carbonyl oxy-2-methylpropane | |
| 2-{[({[(tert-butoxy)carbonyl]amino}oxy)carbonyl]oxy}-2-methylpropane |
Specifications
| 85006-25-3 | |
| White | |
| MFCD00034797 | |
| tert-butyl tert-butoxycarbonyl oxycarbamate, n,o-di-boc-hydroxylamine, tert-butyl n-tert-butoxycarbonyloxy carbamate, n,o-bis tert-butoxycarbonyl hydroxylamine, tert-butyl tert-butoxycarbonyl oxy carbamate, tert-butyl 1,1-dimethylethoxy carbonyl oxy carbamate, tert-butyl tert-butoxycarbonyloxy carbamate, tert-butoxycarbonylamino tert-butyl carbonate, tert-butyl 2-methylpropan-2-yl oxycarbonylamino carbonate, 2-tert-butoxy carbonyl amino oxy carbonyl oxy-2-methylpropane | |
| CC(C)(C)OC(=O)NOC(=O)OC(C)(C)C | |
| 233.26 | |
| 233.26 | |
| Crystalline Powder |
| 69°C | |
| C10H19NO5 | |
| 1 g | |
| AGOSGCWATIJZHQ-UHFFFAOYSA-N | |
| 2-{[({[(tert-butoxy)carbonyl]amino}oxy)carbonyl]oxy}-2-methylpropane | |
| 2728863 | |
| ≥98.0% (GC) | |
| N,O-Bis(tert-butoxycarbonyl)hydroxylamine |
Safety and Handling
TSCA : No
Recommended Storage : Freezer