missing translation for 'onlineSavingsMsg'
Learn More

N,O-Bis(tert-butoxycarbonyl)hydroxylamine 98.0+%, TCI America™

Catalog No. B47805G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B47805G 5 g
B47801G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B47805G Supplier TCI America Supplier No. B47805G
Only null left

Chemical Identifiers

CAS 85006-25-3
Molecular Formula C10H19NO5
Molecular Weight (g/mol) 233.26
MDL Number MFCD00034797
InChI Key AGOSGCWATIJZHQ-UHFFFAOYSA-N
Synonym tert-butyl tert-butoxycarbonyl oxycarbamate, n,o-di-boc-hydroxylamine, tert-butyl n-tert-butoxycarbonyloxy carbamate, n,o-bis tert-butoxycarbonyl hydroxylamine, tert-butyl tert-butoxycarbonyl oxy carbamate, tert-butyl 1,1-dimethylethoxy carbonyl oxy carbamate, tert-butyl tert-butoxycarbonyloxy carbamate, tert-butoxycarbonylamino tert-butyl carbonate, tert-butyl 2-methylpropan-2-yl oxycarbonylamino carbonate, 2-tert-butoxy carbonyl amino oxy carbonyl oxy-2-methylpropane
PubChem CID 2728863
IUPAC Name 2-{[({[(tert-butoxy)carbonyl]amino}oxy)carbonyl]oxy}-2-methylpropane
SMILES CC(C)(C)OC(=O)NOC(=O)OC(C)(C)C

Specifications

Melting Point 69°C
Color White
Quantity 5 g
Formula Weight 233.26
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N,O-Bis(tert-butoxycarbonyl)hydroxylamine
TSCA No
Recommended Storage Freezer