missing translation for 'onlineSavingsMsg'
Learn More

n-Nonylbenzene, 97%

Catalog No. p-7020859
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1295709 10 g
AAA1295718 50 g
AAA1295730 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1295709 Supplier Thermo Scientific Chemicals Supplier No. A1295709

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1081-77-2 | C15H24 | 204.357 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1081-77-2
Molecular Formula C15H24
Molecular Weight (g/mol) 204.357
MDL Number MFCD00009575
InChI Key LIXVMPBOGDCSRM-UHFFFAOYSA-N
Synonym 1-phenylnonane, benzene, nonyl, n-nonylbenzene, nonane, 1-phenyl, 1-nonylbenzene, unii-58ak68ov26, benzene, c6-12-alkyl derivs., n-nonyl benzene, phenyl-nonane, c6-c12 alkylbenzene
PubChem CID 14126
IUPAC Name nonylbenzene
SMILES CCCCCCCCCC1=CC=CC=C1

Specifications

Density 0.858
Boiling Point 281°C to 283°C
Flash Point >110°C (230°F)
Refractive Index 1.483
Quantity 10 g
Beilstein 1907173
Formula Weight 204.36
Percent Purity 97%
Chemical Name or Material n-Nonylbenzene
EINECSNumber 214-103-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only