missing translation for 'onlineSavingsMsg'
Learn More

n-Nonadecane, 99%

Catalog No. AAL0570122
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0570122 100 g
AAL0570114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0570122 Supplier Thermo Scientific Chemicals Supplier No. L0570122
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 629-92-5 | C19H40 | 268.529 g/mol

n-Nonadecane is used as a fragrance agent in cosmetics and perfumes. It is used by egg parasitoid Trissolcus basalis to discriminate between female and male hosts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
n-Nonadecane is used as a fragrance agent in cosmetics and perfumes. It is used by egg parasitoid Trissolcus basalis to discriminate between female and male hosts.

Solubility
Soluble in Chloroform, Hexane.

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in well sealed container.

Chemical Identifiers

CAS 629-92-5
Molecular Formula C19H40
Molecular Weight (g/mol) 268.529
MDL Number MFCD00009012
InChI Key LQERIDTXQFOHKA-UHFFFAOYSA-N
Synonym n-nonadecane, nonadekan, unii-nmy21d3y5t, nmy21d3y5t, unknown branched fragment of phospholipid, ch3-ch2 17-ch3, unknown phospholipid fragment, nonadecane, analytical standard, upl
PubChem CID 12401
ChEBI CHEBI:32927
IUPAC Name nonadecane
SMILES CCCCCCCCCCCCCCCCCCC

Specifications

Melting Point 32°C to 34°C
Density 0.786
Boiling Point 330°C to 331°C
Flash Point 168°C (334°F)
Refractive Index 1.441
Quantity 100 g
Beilstein 1742892
Solubility Information Soluble in Chloroform,Hexane.
Formula Weight 268.53
Percent Purity 99%
Chemical Name or Material n-Nonadecane
Show More Show Less
Hazard Category H304
Precautionary Statement P301+P310-P331-P501c
EINECSNumber 211-116-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only