missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-n-Octyl-3,4-thiophenedicarboximide 98.0+%, TCI America™
Supplier: TCI America O03941G
Specifications
| N-n-Octyl-3,4-thiophenedicarboximide | |
| 124°C | |
| C14H19NO2S | |
| 1 g | |
| QMNVUZQWXKLEFP-UHFFFAOYSA-N | |
| 5-octyl-4H,5H,6H-thieno[3,4-c]pyrrole-4,6-dione | |
| 67122608 | |
| ≥98.0% (GC) |
| 773881-43-9 | |
| White | |
| MFCD23703118 | |
| 5-n-Octyl-4H-thieno[3,4-c]pyrrole-4,6(5H)-dione | |
| CCCCCCCCN1C(=O)C2=CSC=C2C1=O | |
| 265.37 | |
| 265.37 | |
| Crystalline Powder |
Chemical Identifiers
| 773881-43-9 | |
| 265.37 | |
| QMNVUZQWXKLEFP-UHFFFAOYSA-N | |
| 67122608 | |
| CCCCCCCCN1C(=O)C2=CSC=C2C1=O |
| C14H19NO2S | |
| MFCD23703118 | |
| 5-n-Octyl-4H-thieno[3,4-c]pyrrole-4,6(5H)-dione | |
| 5-octyl-4H,5H,6H-thieno[3,4-c]pyrrole-4,6-dione |
Safety and Handling
TSCA : No