missing translation for 'onlineSavingsMsg'
Learn More

N-n-Octyl-3,4-thiophenedicarboximide 98.0+%, TCI America™

Catalog No. O0394200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O0394200MG 200 mg
O03941G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O0394200MG Supplier TCI America Supplier No. O0394200MG
Only null left

Chemical Identifiers

CAS 773881-43-9
Molecular Formula C14H19NO2S
Molecular Weight (g/mol) 265.37
MDL Number MFCD23703118
InChI Key QMNVUZQWXKLEFP-UHFFFAOYSA-N
Synonym 5-n-Octyl-4H-thieno[3,4-c]pyrrole-4,6(5H)-dione
PubChem CID 67122608
IUPAC Name 5-octyl-4H,5H,6H-thieno[3,4-c]pyrrole-4,6-dione
SMILES CCCCCCCCN1C(=O)C2=CSC=C2C1=O

Specifications

Melting Point 124°C
Color White
Quantity 200 mg
Formula Weight 265.37
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-n-Octyl-3,4-thiophenedicarboximide