missing translation for 'onlineSavingsMsg'
Learn More

N,N,O-Triacetylhydroxylamine 99.0+%, TCI America™

Catalog No. T120225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T120225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T120225G Supplier TCI America Supplier No. T120225G
Only null left

Chemical Identifiers

CAS 17720-63-7
Molecular Formula C6H9NO4
Molecular Weight (g/mol) 159.14
MDL Number MFCD00060099
InChI Key NGULNJDDRUSDPR-UHFFFAOYSA-N
PubChem CID 536072
IUPAC Name N-acetylacetamido acetate
SMILES CC(=O)ON(C(C)=O)C(C)=O

Specifications

Color Yellow
Boiling Point 216°C
Quantity 25 g
Formula Weight 159.14
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material N,N,O-Triacetylhydroxylamine