missing translation for 'onlineSavingsMsg'
Learn More

N,N',N″-Trimethyldiethylenetriamine 95.0+%, TCI America™

Catalog No. B43045ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B43045ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B43045ML Supplier TCI America Supplier No. B43045ML

Chemical Identifiers

CAS 105-84-0
Molecular Formula C7H19N3
Molecular Weight (g/mol) 145.25
MDL Number MFCD00025637
InChI Key ODZZIKZQNODXFS-UHFFFAOYSA-N
Synonym 1,4,7-trimethyldiethylenetriamine, n1,n2-dimethyl-n1-2-methylamino ethyl ethane-1,2-diamine, n,n',n-trimethyldiethylenetriamine, 1,2-ethanediamine, n,n'-dimethyl-n-2-methylamino ethyl, n,n'-dimethyl-n-2-methylamino ethyl ethylenediamine, methyl 2-methyl 2-methylamino ethyl amino ethyl amine, 1,2-ethanediamine, n1,n2-dimethyl-n1-2-methylamino ethyl
PubChem CID 66930
IUPAC Name N,N'-dimethyl-N'-[2-(methylamino)ethyl]ethane-1,2-diamine
SMILES CNCCN(C)CCNC

Specifications

Color Yellow
Boiling Point 56°C
Quantity 5 mL
UN Number 2735
Formula Weight 145.25
Percent Purity ≥95.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N,N′,N′′-Trimethyldiethylenetriamine