missing translation for 'onlineSavingsMsg'
Learn More

N,N,N'-Triethylethylenediamine 98.0+%, TCI America™

$113.71 - $352.30

Chemical Identifiers

CAS 105-04-4
Molecular Formula C8H20N2
Molecular Weight (g/mol) 144.26
MDL Number MFCD00009054
InChI Key HDCAZTXEZQWTIJ-UHFFFAOYSA-N
Synonym n,n,n'-triethylethylenediamine, triethylethylenediamine, n,n,n'-triethylethane-1,2-diamine, 1,2-ethanediamine, n,n,n'-triethyl, 2-diethylamino ethyl ethyl amine, 1,2-ethanediamine, n1,n1,n2-triethyl, ethylenediamine, n,n,n'-triethyl, n1,n1,n2-triethyl-1,2-ethanediamine, n,n,n-triethylenediamine, diethyl 2-ethylaminoethyl amine
PubChem CID 66040
IUPAC Name [2-(diethylamino)ethyl](ethyl)amine
SMILES CCNCCN(CC)CC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
T07435ML
View Documents
TCI America
T07435ML
5 mL N/A
Only null left
 
T074325ML
View Documents
TCI America
T074325ML
25 mL N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

105-04-4
144.26
HDCAZTXEZQWTIJ-UHFFFAOYSA-N
66040
CCNCCN(CC)CC
C8H20N2
MFCD00009054
n,n,n'-triethylethylenediamine, triethylethylenediamine, n,n,n'-triethylethane-1,2-diamine, 1,2-ethanediamine, n,n,n'-triethyl, 2-diethylamino ethyl ethyl amine, 1,2-ethanediamine, n1,n1,n2-triethyl, ethylenediamine, n,n,n'-triethyl, n1,n1,n2-triethyl-1,2-ethanediamine, n,n,n-triethylenediamine, diethyl 2-ethylaminoethyl amine
[2-(diethylamino)ethyl](ethyl)amine

Specifications

105-04-4
145°C
MFCD00009054
2734
HDCAZTXEZQWTIJ-UHFFFAOYSA-N
[2-(diethylamino)ethyl](ethyl)amine
66040
≥98.0% (GC,T)
N,N,N′-Triethylethylenediamine
Yellow
C8H20N2
5 mL
n,n,n'-triethylethylenediamine, triethylethylenediamine, n,n,n'-triethylethane-1,2-diamine, 1,2-ethanediamine, n,n,n'-triethyl, 2-diethylamino ethyl ethyl amine, 1,2-ethanediamine, n1,n1,n2-triethyl, ethylenediamine, n,n,n'-triethyl, n1,n1,n2-triethyl-1,2-ethanediamine, n,n,n-triethylenediamine, diethyl 2-ethylaminoethyl amine
CCNCCN(CC)CC
144.26
144.26
Liquid
Videos
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'