missing translation for 'onlineSavingsMsg'
Learn More

N,N,N'-Triethylethylenediamine 98.0+%, TCI America™

Catalog No. T07435ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T07435ML 5 mL
T074325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T07435ML Supplier TCI America Supplier No. T07435ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 105-04-4
Molecular Formula C8H20N2
Molecular Weight (g/mol) 144.26
MDL Number MFCD00009054
InChI Key HDCAZTXEZQWTIJ-UHFFFAOYSA-N
Synonym n,n,n'-triethylethylenediamine, triethylethylenediamine, n,n,n'-triethylethane-1,2-diamine, 1,2-ethanediamine, n,n,n'-triethyl, 2-diethylamino ethyl ethyl amine, 1,2-ethanediamine, n1,n1,n2-triethyl, ethylenediamine, n,n,n'-triethyl, n1,n1,n2-triethyl-1,2-ethanediamine, n,n,n-triethylenediamine, diethyl 2-ethylaminoethyl amine
PubChem CID 66040
IUPAC Name [2-(diethylamino)ethyl](ethyl)amine
SMILES CCNCCN(CC)CC

Specifications

Color Yellow
Boiling Point 145°C
Quantity 5 mL
UN Number 2734
Formula Weight 144.26
Percent Purity ≥98.0% (GC,T)
Grade Not Available
Physical Form Liquid
Chemical Name or Material N,N,N′-Triethylethylenediamine