missing translation for 'onlineSavingsMsg'
Learn More

N,N,N',N'-Tetrapropylmalonamide 97.0+%, TCI America™

Catalog No. T19675G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T19675G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T19675G Supplier TCI America Supplier No. T19675G
Only null left

Chemical Identifiers

CAS 143356-43-8
Molecular Formula C15H30N2O2
Molecular Weight (g/mol) 270.417
MDL Number MFCD03964596
InChI Key GXMHNVKZIWPXAP-UHFFFAOYSA-N
PubChem CID 2384358
IUPAC Name N,N,N',N'-tetrapropylpropanediamide
SMILES CCCN(CCC)C(=O)CC(=O)N(CCC)CCC

Specifications

Color Yellow
Quantity 5 g
Formula Weight 270.42
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material N,N,N′,N′-Tetrapropylmalonamide