missing translation for 'onlineSavingsMsg'
Learn More

N,N,N',N'-Tetraphenylbenzidine (purified by sublimation) 99.0+%, TCI America™

Catalog No. T32661G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T32661G 1 g
T32665G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T32661G Supplier TCI America Supplier No. T32661G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 15546-43-7
Molecular Formula C36H28N2
Molecular Weight (g/mol) 488.63
MDL Number MFCD00228123
InChI Key DCZNSJVFOQPSRV-UHFFFAOYSA-N
Synonym n,n,n',n'-tetraphenylbenzidine, tetraphenylbenzidine, n4,n4,n4',n4'-tetraphenyl-1,1'-biphenyl-4,4'-diamine, n,n,n,n-tetraphenylbenzidine, 1,1'-biphenyl-4,4'-diamine, n,n,n',n'-tetraphenyl, n,n'-diphenyl-n,n',di-m-tolyl-benzidine, n,n,n',n'-tetraphenyl-1,1'-biphenyl-4,4'-diamine, 4,4'-bis diphenylamino biphenyl, 1,1'-biphenyl-4,4'-diamine, n4,n4,n4',n4'-tetraphenyl
PubChem CID 84981
IUPAC Name N4,N4,N4',N4'-tetraphenyl-[1,1'-biphenyl]-4,4'-diamine
SMILES C1=CC=C(C=C1)N(C1=CC=CC=C1)C1=CC=C(C=C1)C1=CC=C(C=C1)N(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 233°C
Color White-Yellow
Quantity 1 g
Formula Weight 488.63
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N,N,N′,N′-Tetraphenylbenzidine (purified by sublimation)