Learn More
N,N,N',N'-Tetramethylmethylenediamine, 99%
CAS: 51-80-9 | C5H14N2 | 102.18 g/mol
Supplier: Thermo Scientific Chemicals L0403722
Description
N,N,N',N'-Tetramethylmethylenediamine is a reagent used as a convenient source of the Mannich intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN,N,N′,N′-Tetramethylmethylenediamine is a reagent used as a convenient source of the Mannich intermediate.
Solubility
Fully miscible in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications
| N,N,N',N'-Tetramethylmethylenediamine | |
| -55°C | |
| 83°C to 85°C | |
| C5H14N2 | |
| (CH3)2NCH2N(CH3)2 | |
| 100 g | |
| 1731946 | |
| Fully miscible in water. | |
| CN(C)CN(C)C | |
| 102.18 | |
| 102.18 |
| 51-80-9 | |
| 0.749 | |
| −12°C (10°F) | |
| 1.401 | |
| MFCD00008328 | |
| UN2733 | |
| n,n,n',n'-tetramethyldiaminomethane, bis dimethylamino methane, methanediamine, n,n,n',n'-tetramethyl, tetramethylmethylenediamine, methylenebis dimethylamine, n,n,n',n'-tetramethylmethylenediamine, unii-z870i525ks, methylenediamine, n,n,n',n'-tetramethyl, dimethylamino methyl dimethylamine, dimethyl dimethylamino methyl amine | |
| VGIVLIHKENZQHQ-UHFFFAOYSA-N | |
| [(dimethylamino)methyl]dimethylamine | |
| 5829 | |
| 99% |
Chemical Identifiers
| 51-80-9 | |
| 102.18 | |
| VGIVLIHKENZQHQ-UHFFFAOYSA-N | |
| 5829 |
| C5H14N2 | |
| MFCD00008328 | |
| n,n,n',n'-tetramethyldiaminomethane, bis dimethylamino methane, methanediamine, n,n,n',n'-tetramethyl, tetramethylmethylenediamine, methylenebis dimethylamine, n,n,n',n'-tetramethylmethylenediamine, unii-z870i525ks, methylenediamine, n,n,n',n'-tetramethyl, dimethylamino methyl dimethylamine, dimethyl dimethylamino methyl amine | |
| CN(C)CN(C)C |
Safety and Handling
GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H225-H314-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
EINECSNumber : 200-124-7
RTECSNumber : PA6700000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only