missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich N,N,N',N'-Tetramethylethylenediamine

Catalog No. T225005ML
Encompass
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T225005ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T225005ML Supplier Sigma Aldrich Supplier No. T225005ML
Only null left

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

N,N,N',N'-Tetramethylethylenediamine

Specifications

CAS 110-18-9
Melting Point -55°C
Density 0.775 g/mL at 20°C
Boiling Point 120°C to 122°C
Molecular Formula C6H16N2
Refractive Index n20/D 1.4179
Linear Formula (CH3)2NCH2CH2N(CH3)2
MDL Number MFCD00008335
Quantity 5 mL
Synonym 1,2-Bis(dimethylamino)ethane; TEMED; TMEDA
Molecular Weight (g/mol) 116.2 g/mol
Show More Show Less