missing translation for 'onlineSavingsMsg'
Learn More

N,N,N',N'-Tetramethylbenzidine 98.0+%, TCI America™

Catalog No. T01415G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T01415G 5 g
T01411G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T01415G Supplier TCI America Supplier No. T01415G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 366-29-0
Molecular Formula C16H20N2
Molecular Weight (g/mol) 240.35
MDL Number MFCD00008310
InChI Key YRNWIFYIFSBPAU-UHFFFAOYSA-N
Synonym n,n,n',n'-tetramethylbenzidine, benzidine, n,n,n',n'-tetramethyl, 4,4'-bis n,n-dimethylamino biphenyl, n,n,n',n'-tetramethyl-p,p'-benzidine, ccris 1000, 1,1'-biphenyl-4,4'-diamine, n,n,n',n'-tetramethyl, n,n,n',n'-tetramethyl-1,1'-biphenyl-4,4'-diamine, 4-4-dimethylamino phenyl phenyl dimethylamine, 1,1'-biphenyl-4,4'-diamine, n4,n4,n4',n4'-tetramethyl
PubChem CID 9702
IUPAC Name N4,N4,N4',N4'-tetramethyl-[1,1'-biphenyl]-4,4'-diamine
SMILES CN(C)C1=CC=C(C=C1)C1=CC=C(C=C1)N(C)C

Specifications

Melting Point 193°C
Color Yellow
Quantity 5 g
Formula Weight 240.35
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material N,N,N′,N′-Tetramethylbenzidine