missing translation for 'onlineSavingsMsg'
Learn More

N,N,N',N'-Tetramethyl-p-phenylenediamine, 98+%

Catalog No. AAL0055909
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0055909 10 g
AAL0055918 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0055909 Supplier Thermo Scientific Chemicals Supplier No. L0055909
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 100-22-1 | C10H16N2 | 164.252 g/mol

N,N,N',N'-Tetramethyl-p-phenylenediamine is a reducing co-substrate for heme peroxidases. It is used for detection of peroxidases on polyacrylamide gels.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N,N′,N′-Tetramethyl-p-phenylenediamine is a reducing co-substrate for heme peroxidases. It is used for detection of peroxidases on polyacrylamide gels.

Solubility
Slightly soluble in cold, more in hot water.

Notes
Air & Light Sensitive. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 100-22-1
Molecular Formula C10H16N2
Molecular Weight (g/mol) 164.252
MDL Number MFCD00008309
InChI Key CJAOGUFAAWZWNI-UHFFFAOYSA-N
Synonym wurster's blue, wurster's reagent, n,n,n',n'-tetramethyl-p-phenylenediamine, n,n,n',n'-tetramethyl-1,4-phenylenediamine, tetramethyl-p-phenylenediamine, 1,4-bis dimethylamino benzene, p-bis dimethylamino benzene, 1,4-benzenediamine, n,n,n',n'-tetramethyl, benzene, 1,4-bis dimethylamino, unii-p4p3ac32zb
PubChem CID 7490
IUPAC Name 1-N,1-N,4-N,4-N-tetramethylbenzene-1,4-diamine
SMILES CN(C)C1=CC=C(C=C1)N(C)C

Specifications

Melting Point 48°C to 52°C
Boiling Point 260°C
Quantity 10 g
UN Number UN1673
Beilstein 1564025
Sensitivity Air and light sensitive
Merck Index 14,9228
Solubility Information Slightly soluble in cold,more in hot water.
Formula Weight 164.25
Percent Purity ≥98%
Chemical Name or Material N,N,N',N'-Tetramethyl-p-phenylenediamine
Show More Show Less
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: PHENYLENEDIAMINES
EINECSNumber 202-831-6
RTECSNumber ST4200000
TSCA Yes
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only