missing translation for 'onlineSavingsMsg'
Learn More

N,N,N',N'-Tetramethyl-O-(N-succinimidyl)uronium Tetrafluoroborate 98.0+%, TCI America™

Catalog No. p-7137121
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T22241G 1 g
T22245G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T22241G Supplier TCI America Supplier No. T22241G
Only null left

Chemical Identifiers

CAS 105832-38-0
Molecular Formula C9H16BF4N3O3
Molecular Weight (g/mol) 301.05
MDL Number MFCD00077875
InChI Key YEBLHMRPZHNTEK-UHFFFAOYSA-N
Synonym tstu, o-n-succinimidyl-1,1,3,3-tetramethyluronium tetrafluoroborate, 2-2,5-dioxopyrrolidin-1-yl-1,1,3,3-tetramethylisouronium tetrafluoroborate, 2-succinimido-1,1,3,3-tetramethyluronium tetrafluoroborate, n,n,n',n'-tetramethyl-o-n-succinimidyl uronium tetrafluoroborate, o-n-succinimidyl-n,n,n',n'-tetramethyluronium tetrafluoroborate, o-n-succinimidyl-n,n,n',n'-tetramethyl-uronium tetrafluoroborate, dimethylamino 2,5-dioxopyrrolidin-1-yl oxy methylidene dimethylazanium tetrafluoroborate, o-n-succinimidyl-n,n,n',n'-tetramethyl uronium bf4, pubchem12781
PubChem CID 9857522
IUPAC Name [(dimethylamino)[(2,5-dioxopyrrolidin-1-yl)oxy]methylidene]dimethylazanium; tetrafluoroboranuide
SMILES F[B-](F)(F)F.CN(C)C(ON1C(=O)CCC1=O)=[N+](C)C

Specifications

Melting Point 195°C
Color White
Quantity 1 g
UN Number 1759
Formula Weight 301.05
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N,N,N′,N′-Tetramethyl-O-(N-succinimidyl)uronium Tetrafluoroborate
TSCA No
Recommended Storage Refrigerator