missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N,N',N'-Tetramethyl-O-(N-succinimidyl)uronium Hexafluorophosphate 98.0+%, TCI America™
Supplier: TCI America T292925G
Specifications
| N,N,N′,N′-Tetramethyl-O-(N-succinimidyl)uronium Hexafluorophosphate | |
| 230°C | |
| C9H16F6N3O3P | |
| 25 g | |
| STWZCCVNXFLDDD-UHFFFAOYSA-N | |
| [(dimethylamino)[(2,5-dioxopyrrolidin-1-yl)oxy]methylidene]dimethylazanium; hexafluoro-λâµ-phosphanuide | |
| 16211151 | |
| ≥98.0% (HPLC,N) |
| 265651-18-1 | |
| White | |
| MFCD01863753 | |
| hstu, n,n,n',n'-tetramethyl-o-n-succinimidyl uronium hexafluorophosphate, o-n-succinimidyl-n,n,n',n'-tetramethyluronium hexafluorophosphate, hstu n,n,n',n'-tetramethyl-o-n-succinimidyl uronium hexafluorophosphate, dimethylamino 2,5-dioxopyrrolidin-1-yl oxy methylidene dimethylazanium hexafluorophosphate, ambotzrl-1039, o-n-succinimidyl-1,1,3,3-tetramethyluronium hexafluorophosphate, 2-succinimido-1,1,3,3-tetramethyluronim hexafluorophosphate, 2-succinimido-1,1,3,3-tetramethyluronium hexafluorophosphate, n,n,n',n'-tetramethyl-o-n-succinimidyl uroniumhexafluorophosphate | |
| F[P-](F)(F)(F)(F)F.CN(C)C(ON1C(=O)CCC1=O)=[N+](C)C | |
| 359.21 | |
| 359.21 | |
| Crystalline Powder |
Chemical Identifiers
| 265651-18-1 | |
| 359.21 | |
| STWZCCVNXFLDDD-UHFFFAOYSA-N | |
| 16211151 | |
| F[P-](F)(F)(F)(F)F.CN(C)C(ON1C(=O)CCC1=O)=[N+](C)C |
| C9H16F6N3O3P | |
| MFCD01863753 | |
| hstu, n,n,n',n'-tetramethyl-o-n-succinimidyl uronium hexafluorophosphate, o-n-succinimidyl-n,n,n',n'-tetramethyluronium hexafluorophosphate, hstu n,n,n',n'-tetramethyl-o-n-succinimidyl uronium hexafluorophosphate, dimethylamino 2,5-dioxopyrrolidin-1-yl oxy methylidene dimethylazanium hexafluorophosphate, ambotzrl-1039, o-n-succinimidyl-1,1,3,3-tetramethyluronium hexafluorophosphate, 2-succinimido-1,1,3,3-tetramethyluronim hexafluorophosphate, 2-succinimido-1,1,3,3-tetramethyluronium hexafluorophosphate, n,n,n',n'-tetramethyl-o-n-succinimidyl uroniumhexafluorophosphate | |
| [(dimethylamino)[(2,5-dioxopyrrolidin-1-yl)oxy]methylidene]dimethylazanium; hexafluoro-λâµ-phosphanuide |
Safety and Handling
TSCA : No
Recommended Storage : Freezer