missing translation for 'onlineSavingsMsg'
Learn More

(+)-N,N,N',N'-Tetramethyl-L-tartardiamide 98.0+%, TCI America™

Catalog No. T18395G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T18395G 5 g
T183925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T18395G Supplier TCI America Supplier No. T18395G
Only null left

Chemical Identifiers

CAS 26549-65-5
Molecular Formula C8H16N2O4
Molecular Weight (g/mol) 204.226
MDL Number MFCD00025672
InChI Key PCYDYHRBODKVEL-PHDIDXHHSA-N
PubChem CID 6610750
IUPAC Name (2R,3R)-2,3-dihydroxy-N,N,N',N'-tetramethylbutanediamide
SMILES CN(C)C(=O)C(C(C(=O)N(C)C)O)O

Specifications

Melting Point 187°C
Color White
Quantity 5 g
Synonym (R,R)-(+)-2,3-Dihydroxy-N,N,N′C,N′C-tetramethylsuccinamide
Formula Weight 204.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (+)-N,N,N′,N′-Tetramethyl-L-tartardiamide