missing translation for 'onlineSavingsMsg'
Learn More

N,N,N',N'-Tetramethyl-1,6-diaminohexane 98.0+%, TCI America™

Catalog No. p-7136333
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T053725ML 25 mL
T0537500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T053725ML Supplier TCI America Supplier No. T053725ML
Only null left

Chemical Identifiers

CAS 111-18-2
Molecular Formula C10H24N2
Molecular Weight (g/mol) 172.316
MDL Number MFCD00008339
InChI Key TXXWBTOATXBWDR-UHFFFAOYSA-N
Synonym n,n,n',n'-tetramethyl-1,6-hexanediamine, 1,6-bis dimethylamino hexane, hexamethylenebis dimethylamine, toyocat mr, koolizer 1, minico tmhd, 1,6-hexanediamine, n,n,n',n'-tetramethyl, n,n,n',n'-tetramethyl-1,6-diaminohexane, n1,n1,n6,n6-tetramethylhexane-1,6-diamine, niax catalyst c-210
PubChem CID 8097
IUPAC Name N,N,N',N'-tetramethylhexane-1,6-diamine
SMILES CN(C)CCCCCCN(C)C

Specifications

Color Colorless
pH 12.1 (5% H2O soln.)
Boiling Point 210°C
Quantity 25 mL
UN Number 2922
Formula Weight 172.32
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N,N,N′,N′-Tetramethyl-1,6-diaminohexane
EINECSNumber (2)-0156
RTECSNumber MO1370000
TSCA Yes