missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N,N',N'-Tetramethyl-1,4-diaminobutane 98.0+%, TCI America™
Specifications
Specifications
| Color | Colorless |
| Boiling Point | 100°C |
| Quantity | 25 mL |
| UN Number | 2734 |
| Formula Weight | 144.26 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Liquid |
| Chemical Name or Material | N,N,N′,N′-Tetramethyl-1,4-diaminobutane |
Safety and Handling
| RTECSNumber | EJ7530000 |
| TSCA | Yes |